Thermo Scientific Chemicals

2,2,2-Trifluoroethanol, 99.8%, extra pure, Thermo Scientific Chemicals

Catalog number: 139755000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroethanol, 99.8%, extra pure, Thermo Scientific Chemicals

Catalog number: 139755000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 139755000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-89-8
IUPAC Name
2,2,2-trifluoroethan-1-ol
Molecular Formula
C2H3F3O
InChI Key
RHQDFWAXVIIEBN-UHFFFAOYSA-N
SMILES
OCC(F)(F)F
Infrared spectrum
Conforms
GC
>=99.75 %
Water
=<0.1 % (K.F.)
pH
5.0 to 7.5 (10 % v/v solution)
Residue after evaporation
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text