Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: 139790050
5 g, Each
Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: 139790050
5 g, Each
Quantity
Catalog number: 139790050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
480-66-0
IUPAC Name
1-(2,4,6-trihydroxyphenyl)ethan-1-one
Molecular Formula
C8H8O4
InChI Key
XLEYFDVVXLMULC-UHFFFAOYSA-N
SMILES
CC(=O)C1=C(O)C=C(O)C=C1O
Appearance (Color)
Yellow to orange to tan
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
217°C to 222°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text