2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific Chemicals
2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 139791000
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS480-66-0
IUPAC Name1-(2,4,6-trihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O4
InChI KeyXLEYFDVVXLMULC-UHFFFAOYSA-N
SMILESCC(=O)C1=C(O)C=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to tan
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point217°C to 222°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only