Thermo Scientific Chemicals

4',5,7-Trihydroxyflavanone, 97%, Thermo Scientific Chemicals

Catalog number: 139820100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

4',5,7-Trihydroxyflavanone, 97%, Thermo Scientific Chemicals

Catalog number: 139820100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139820100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
480-41-1
IUPAC Name
5,7-dihydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Molecular Formula
C15H12O5
InChI Key
FTVWIRXFELQLPI-UHFFFAOYNA-N
SMILES
OC1=CC=C(C=C1)C1CC(=O)C2=C(O)C=C(O)C=C2O1
HPLC
>=96.0 %
Melting point
247°C to 254°C (Dec)
Appearance (Color)
Beige to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text