Thermo Scientific Chemicals

3,4,5-Trimethoxyaniline, 97%, Thermo Scientific Chemicals

Catalog number: 139900100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxyaniline, 97%, Thermo Scientific Chemicals

Catalog number: 139900100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139900100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
24313-88-0
IUPAC Name
3,4,5-trimethoxyaniline
Molecular Formula
C9H13NO3
InChI Key
XEFRNCLPPFDWAC-UHFFFAOYSA-N
SMILES
COC1=CC(N)=CC(OC)=C1OC
Infrared spectrum
Conforms
Melting point
108°C to 113°C
Appearance (Color)
Off-white to beige to light brown
Appearance (Form)
Powder
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text