Thermo Scientific Chemicals

3,4,5-Trimethoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 139931000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 139931000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 139931000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
118-41-2
IUPAC Name
3,4,5-trimethoxybenzoic acid
Molecular Formula
C10H12O5
InChI Key
SJSOFNCYXJUNBT-UHFFFAOYSA-N
SMILES
COC1=CC(=CC(OC)=C1OC)C(O)=O
Infrared spectrum
Conforms
Appearance (Color)
White to beige
Melting point
168°C to 172°C
Appearance (Form)
Crystalline powder
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text