Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals
Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number 140052500
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.00 
Save 6.35 (14%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.00 
Save 6.35 (14%)
Add to cart
Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals
Catalog number140052500
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.00 
Save 6.35 (14%)
-
Add to cart
Chemical Identifiers
CAS3282-30-2
IUPAC Name2,2-dimethylpropanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyJVSFQJZRHXAUGT-UHFFFAOYSA-N
SMILESCC(C)(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOCl98.5 to 101.5 %
Appearance (Color)Clear colorless to light pink
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4110 to 1.4130 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only