Thermo Scientific Chemicals

Trimethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 140071000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Trimethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 140071000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 140071000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
593-81-7
IUPAC Name
hydrogen trimethylamine chloride
Molecular Formula
C3H10ClN
InChI Key
SZYJELPVAFJOGJ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)C
Appearance (Color)
White to cream
Titration with HClO4
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text