Trimethyl borate, 99%, Thermo Scientific Chemicals
Trimethyl borate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyl borate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 140121000
Price (USD)/ Each
19.65
Online exclusive
22.50 
Save 2.85 (13%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
19.65
Online exclusive
22.50 
Save 2.85 (13%)
Add to cart
Trimethyl borate, 99%, Thermo Scientific Chemicals
Catalog number140121000
Price (USD)/ Each
19.65
Online exclusive
22.50 
Save 2.85 (13%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrimethyl borate
Molecular FormulaC3H9BO3
InChI KeyWRECIMRULFAWHA-UHFFFAOYSA-N
SMILESCOB(OC)OC
Molecular Weight (g/mol)103.91
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Refractive index1.3562 to 1.3582 (20°C, 589 nm)
Appearance (Color)Clear colorless

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only