Thermo Scientific Chemicals

Trimethyl borate, 99%, Thermo Scientific Chemicals

Catalog number: 140125000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Trimethyl borate, 99%, Thermo Scientific Chemicals

Catalog number: 140125000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 140125000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-43-7
IUPAC Name
trimethyl borate
Molecular Formula
C3H9BO3
InChI Key
WRECIMRULFAWHA-UHFFFAOYSA-N
SMILES
COB(OC)OC
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %
Refractive index
1.3562 to 1.3582 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text