Thermo Scientific Chemicals

Trimethylhydroquinone, 97%, Thermo Scientific Chemicals

Catalog number: 140155000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Trimethylhydroquinone, 97%, Thermo Scientific Chemicals

Catalog number: 140155000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 140155000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
700-13-0
IUPAC Name
2,3,5-trimethylbenzene-1,4-diol
Molecular Formula
C9H12O2
InChI Key
AUFZRCJENRSRLY-UHFFFAOYSA-N
SMILES
CC1=CC(O)=C(C)C(C)=C1O
Infrared spectrum
Conforms
Appearance (Color)
White to beige to orange
Appearance (Form)
Crystalline powder
Melting point
168°C to 174°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text