Thermo Scientific Chemicals

Trimethylsulfonium Iodide, 98%, Thermo Scientific Chemicals

Catalog number: 140251000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Trimethylsulfonium Iodide, 98%, Thermo Scientific Chemicals

Catalog number: 140251000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 140251000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2181-42-2
IUPAC Name
trimethylsulfanium iodide
Molecular Formula
C3H9IS
InChI Key
VFJYIHQDILEQNR-UHFFFAOYSA-M
SMILES
[I-].C[S+](C)C
Appearance (Color)
White to yellow
Titration Argentometric
>=97.5 %
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text