Thermo Scientific Chemicals

Trimethylsulfoxonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: 140261000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Trimethylsulfoxonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: 140261000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 140261000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1774-47-6
IUPAC Name
trimethyl(oxo)-λ⁶-sulfanylium iodide
Molecular Formula
C3H9IOS
InChI Key
BPLKQGGAXWRFOE-UHFFFAOYSA-M
SMILES
[I-].C[S+](C)(C)=O
Titration Argentometric
>=97.5 %
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text