Tri-n-octylamine, 97%, Thermo Scientific Chemicals
Tri-n-octylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-octylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 140281000
Price (USD)/ Each
67.65
Online exclusive
75.10 
Save 7.45 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
67.65
Online exclusive
75.10 
Save 7.45 (10%)
Add to cart
Tri-n-octylamine, 97%, Thermo Scientific Chemicals
Catalog number140281000
Price (USD)/ Each
67.65
Online exclusive
75.10 
Save 7.45 (10%)
-
Add to cart
Chemical Identifiers
CAS1116-76-3
IUPAC Nametrioctylamine
Molecular FormulaC24H51N
InChI KeyXTAZYLNFDRKIHJ-UHFFFAOYSA-N
SMILESCCCCCCCCN(CCCCCCCC)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4480 to 1.4500 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only