Thermo Scientific Chemicals

Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 140400025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 140400025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 140400025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-83-5
IUPAC Name
(chlorodiphenylmethyl)benzene
Molecular Formula
C19H15Cl
InChI Key
JBWKIWSBJXDJDT-UHFFFAOYSA-N
SMILES
ClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to off-white to yellow to light beige
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
109°C to 113°C
Titration Argentometric
>=97.5 % (ex Cl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text