Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

DPBS, calcium, magnesium

Catalog number: 14040182
Have Questions?
Gibco™

DPBS, calcium, magnesium

Catalog number: 14040182
Have Questions?
Catalog Number
14040182
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1404018210 x 500 mL
-
14040141100 mL
-
14040133500 mL
-
140401171,000 mL
-
140402166 x 1,000 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.


This DPBS is modified as follows:
WithWithout
• Calcium• Phenol Red
• Magnesium 


The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Additive
No Sodium Pyruvate
Classification
Animal Origin-Free
Dilution
1 X
Product Line
Gibco™
Product Type
DPBS
Quantity
100 μg
Solution Type
Dulbecco's Phosphate Buffered Saline
For Use With (Application)
Washing Cells Before Dissociation, Transporting Cells or Tissue Samples, Diluting Cells for Counting, and Preparing Reagents
Green Features
Sustainable packaging
Osmolality
270 - 300 mOsm/kg
pH
7.0 to 7.2
Phenol Red Indicator
No Phenol Red
Shipping Condition
Gel Ice
Sterility
Sterile-filtered
Form
Liquid
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard

Contents & Storage

Storage conditions: 2-8° C
Shelf life: 36 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text