Triphenylmethyl mercaptan, 97%, Thermo Scientific Chemicals
Triphenylmethyl mercaptan, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triphenylmethyl mercaptan, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 140410250
Price (USD)/ Each
77.65
Online exclusive
86.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
77.65
Online exclusive
86.10 
Save 8.45 (10%)
Add to cart
Triphenylmethyl mercaptan, 97%, Thermo Scientific Chemicals
Catalog number140410250
Price (USD)/ Each
77.65
Online exclusive
86.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS3695-77-0
IUPAC Nametriphenylmethanethiol
Molecular FormulaC19H16S
InChI KeyJQZIKLPHXXBMCA-UHFFFAOYSA-N
SMILESSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point102°C to 109°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only