Thermo Scientific Chemicals

Triphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog number: 140422500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Triphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog number: 140422500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 140422500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
603-35-0
IUPAC Name
triphenylphosphane
Molecular Formula
C18H15P
InChI Key
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals or flakes
Infrared spectrum
Conforms
Melting point
78.5°C to 81.5°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text