Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
This product ships via ground from US northeast.
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry

Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 140500025
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
Add to cart
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Catalog number140500025
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 290 nm A: =<0.2
Titration with HClO4>=99.0 %
Heavy metals (as Pb)=<5 ppm
pH10.0 to 11.5 (5 % in water)
UV(40 % in water) 1 cm cell
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only