Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trithiocyanuric acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 140530250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS638-16-4
IUPAC Name1,3,5-triazinane-2,4,6-trithione
Molecular FormulaC3H3N3S3
InChI KeyWZRRRFSJFQTGGB-UHFFFAOYSA-N
SMILESS=C1NC(=S)NC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only