Thermo Scientific Chemicals

DL-Tropic acid, 97%, Thermo Scientific Chemicals

Catalog number: 140550250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Tropic acid, 97%, Thermo Scientific Chemicals

Catalog number: 140550250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 140550250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
552-63-6
IUPAC Name
3-hydroxy-2-phenylpropanoic acid
Molecular Formula
C9H10O3
InChI Key
JACRWUWPXAESPB-UHFFFAOYNA-N
SMILES
OCC(C(O)=O)C1=CC=CC=C1
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
115°C to 120°C
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text