Tropine, 98%, Thermo Scientific Chemicals
Tropine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tropine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 140560250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS120-29-6
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-ol
Molecular FormulaC8H15NO
InChI KeyCYHOMWAPJJPNMW-UHFFFAOYNA-N
SMILESCN1C2CCC1CC(O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dried substance)
Water=<2 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only