Thermo Scientific Chemicals

Tropine, 98%, Thermo Scientific Chemicals

Catalog number: 140561000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tropine, 98%, Thermo Scientific Chemicals

Catalog number: 140561000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 140561000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-29-6
IUPAC Name
8-methyl-8-azabicyclo[3.2.1]octan-3-ol
Molecular Formula
C8H15NO
InChI Key
CYHOMWAPJJPNMW-UHFFFAOYNA-N
SMILES
CN1C2CCC1CC(O)C2
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 % (On dried substance)
Water
=<2 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text