Tropinone, 99%, Thermo Scientific Chemicals
Tropinone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tropinone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 140570050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS532-24-1
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-one
Molecular FormulaC8H13NO
InChI KeyQQXLDOJGLXJCSE-UHFFFAOYNA-N
SMILESCN1C2CCC1CC(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
Infrared spectrumConforms
GC>=98.5 %
Melting point41°C to 44°C
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only