Tropinone, 99%
Tropinone, 99%
Tropinone, 99%
Thermo Scientific Chemicals

Tropinone, 99%

CAS: 532-24-1 | C8H13NO | 139.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 140570100
Price (EUR)
56,53
Online Exclusive
66,50
Save 9,97 (15%)
Each
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
56,53
Online Exclusive
66,50
Save 9,97 (15%)
Each
Add to cart
Chemical Identifiers
CAS532-24-1
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-one
Molecular FormulaC8H13NO
InChI KeyQQXLDOJGLXJCSE-UHFFFAOYNA-N
SMILESCN1C2CCC1CC(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
Infrared spectrumConforms
GC>=98.5 %
Melting point41°C to 44°C
Appearance (Form)Crystalline powder or crystals
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0393504Certificate of AnalysisDec 15, 2022140570050, 140570500, 140570100
A0395436Certificate of AnalysisDec 15, 2022140570050, 140570500, 140570100
A0409968Certificate of AnalysisDec 13, 2022140570050, 140570500, 140570100
A0412878Certificate of AnalysisApr 18, 2022140570050, 140570500, 140570100
4 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H302 - Harmful if swallowed

H314 - Causes severe skin burns and eye damage

Precautionary Statements

P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

P280 - Wear eye protection/ face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection