Tyramine, 97%, Thermo Scientific Chemicals
Tyramine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tyramine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 140610050
Price (USD)/ Each
40.65
Online exclusive
44.50 
Save 3.85 (9%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
44.50 
Save 3.85 (9%)
Add to cart
Tyramine, 97%, Thermo Scientific Chemicals
Catalog number140610050
Price (USD)/ Each
40.65
Online exclusive
44.50 
Save 3.85 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-(2-aminoethyl)phenol
Molecular FormulaC8H11NO
InChI KeyDZGWFCGJZKJUFP-UHFFFAOYSA-N
SMILESNCCC1=CC=C(O)C=C1
Molecular Weight (g/mol)137.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to light brown
Infrared spectrumConforms
Melting point158°C to 163°C
Appearance (Form)Powder
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only