DL-m-Tyrosine, 98.50%
DL-m-Tyrosine, 98.50%
Thermo Scientific Chemicals

DL-m-Tyrosine, 98.50%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 140630010
Price (USD)/ Each
80.80
-
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
80.80
Add to cart
DL-m-Tyrosine, 98.50%
Catalog number140630010
Price (USD)/ Each
80.80
-
Add to cart
Chemical Identifiers
CAS775-06-4
IUPAC Name2-amino-3-(3-hydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO3
InChI KeyJZKXXXDKRQWDET-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC(O)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Titration with HClO4>=98.0 %
Water=<0.5 % (K.F.)
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only