L-Tyrosine methyl ester, 98%
L-Tyrosine methyl ester, 98%
Thermo Scientific Chemicals

L-Tyrosine methyl ester, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 140650050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC10H13NO3
InChI KeyMWZPENIJLUWBSY-VIFPVBQESA-N
SMILESCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Molecular Weight (g/mol)195.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=97.5 %
Specific optical rotation+23° to +29° (20°C, 589 nm) (c=2.4, CH3OH)
Appearance of solution(5% soln. in methanol) Clear to slightly hazy
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only