1x1 image pixel for data collection
Nunc™ Cell-Culture Treated Multidishes
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Cell-Culture Treated Multidishes
Thermo Scientific™

Nunc™ Cell-Culture Treated Multidishes

Nunc Cell-Culture Treated Multidishes receive the proprietary Nunclon Delta surface treatment that promotes maximum adhesion for a broad range of cell types.
Promo ImagePromotion
Promo code:RCELL51
Stock your lab with plasticware essentials so that you can culture with confidence
From now until Dec 31, get 51% off various cell culture plasticware like dishes, flasks, plates and tubes that help achieve reproducible results when you spend $1,000Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per CaseNo. per Pack
140675Promo Image675/Cs.1 Ea.
142475Promo Image2475/Cs.1 Ea.
150628Promo Image1275/Cs.1 Ea.
150687Promo Image4875/Cs.1 Ea.
176740Promo Image4120/Cs.4 Pk.
140685Promo Image685/Cs.5 Pk.
142485Promo Image2485/Cs.5 Pk.
152640Promo Image4885/Cs.5 Pk.
Catalog number 140675
Price (USD)
344.65
Online Exclusive
368.00
Save 23.35 (6%)
Case of 75
Estimated availability date  Pending  
Add to cart
No. of Wells:
6
No. per Case:
75/Cs.
No. per Pack:
1 Ea.
Price (USD)
344.65
Online Exclusive
368.00
Save 23.35 (6%)
Case of 75
Add to cart

Thermo Scientific Nunc Cell-Culture Treated Multidishes receive the Nunclon Delta surface treatment. This proprietary surface modification promotes maximum adhesion for a broad range of cell types and ensures optimum performance.

Nunclon Delta surface treatment ensures optimum performance for cell adhesion.

  • Flat well bottom and stackable
  • Raised well rims lower risk of cross contamination
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Excellent optical quality
  • Alphanumerical codes for well identification

Round well styles

  • Vented lids for gas exchange
  • Recessed areas around wells act as a water reservoir to reduce evaporation from wells
  • Lids with notched corners for easy orientation

Rectangular well styles

  • Large culture area
  • Accept standard glass cover slips: 24 x 60 mm for 4-well plate and 22 x 22 mm for 8-well plate

Additional information

24-well plates are 17 mm deep; 4-well plate is 10 mm deep

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceWith certificate of traceability and quality (one certificate per case).
Culture Area9.6 sq. cm
IncludesLid
Length (Metric)127.76 mm
No. per Case75/Cs.
No. per Pack1 Ea.
PackagingIndividually Packed
Product TypeCell Culture Dish
Volume (Metric) Working3 mL
Width (Metric)85.46 mm
MaterialPolystyrene
No. of Wells6
SterilitySterile
Surface TreatmentNunclon Delta
Volume (Metric) Well3 mL (Well)
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 75

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.

What is Nunclon Delta?

Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link