1-Undecanol, 98%, Thermo Scientific Chemicals
1-Undecanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Undecanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number 140685000
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
Add to cart
1-Undecanol, 98%, Thermo Scientific Chemicals
Catalog number140685000
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS112-42-5
IUPAC Nameundecan-1-ol
Molecular FormulaC11H24O
InChI KeyKJIOQYGWTQBHNH-UHFFFAOYSA-N
SMILESCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4370 to 1.4420 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only