Thermo Scientific Chemicals

Uridine, 99%

Catalog number: 140775000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Uridine, 99%

Catalog number: 140775000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 140775000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
58-96-8
IUPAC Name
1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C9H12N2O6
InChI Key
DRTQHJPVMGBUCF-UHFFFAOYNA-N
SMILES
OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O
UV spectrophotometry
>=98 % (lambda max. 262 nm, pH 7)
Loss on drying
=<1 % (105°C, 2 hrs)
Heavy metals (as Pb)
=<10 ppm
Sulfated ash
=<0.1 %
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text