γ-Valerolactone, 98%, Thermo Scientific Chemicals
γ-Valerolactone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

γ-Valerolactone, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 140795000
Price (USD)/ Each
124.65
Online exclusive
147.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
124.65
Online exclusive
147.00 
Save 22.35 (15%)
Add to cart
γ-Valerolactone, 98%, Thermo Scientific Chemicals
Catalog number140795000
Price (USD)/ Each
124.65
Online exclusive
147.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name5-methyloxolan-2-one
Molecular FormulaC5H8O2
InChI KeyGAEKPEKOJKCEMS-UHFFFAOYNA-N
SMILESCC1CCC(=O)O1
Molecular Weight (g/mol)100.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4310 to 1.4340 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only