DPBS (10X), calcium, magnesium
Save up to 29% on your order of cell culture products. View promotion details >
DPBS (10X), calcium, magnesium
Gibco™

DPBS (10X), calcium, magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Catalog NumberQuantity
14080055500 mL
Catalog number 14080055
Price (USD)
68.50
Each
-
Add to cart
Quantity:
500 mL
Customize this product
Price (USD)
68.50
Each
Add to cart
Ask our AI about this Product

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.


This DPBS is modified as follows:
WithWithout
• Calcium• Phenol Red
• Magnesium 


The complete formulation is available.

Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
AdditiveNo Sodium Pyruvate
ClassificationAnimal Origin-Free
Dilution10 X
For Use With (Application)Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue; Diluting Cells for Counting; and Preparing Reagents
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality2900 - 3150 mOsm/kg
Phenol Red IndicatorNo Phenol Red
Product LineGibco™
Quantity500 mL
Shipping ConditionRoom Temperature
SterilitySterile-filtered
FormLiquid
Product TypeDPBS
Solution TypeDulbecco's Phosphate Buffered Saline
pH4.4 - 4.7
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C.
Shelf life: 24 months from date of manufacture.
four-plex-image-COS7-live-cell zoomed

Follow the 5 steps to publication-quality live-cell imaging.
Download your free guide today ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link