Thermo Scientific Chemicals

L-Valine 99%

Catalog number: 140810025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

L-Valine 99%

Catalog number: 140810025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 140810025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
72-18-4
IUPAC Name
(2S)-2-amino-3-methylbutanoic acid
Molecular Formula
C5H11NO2
InChI Key
KZSNJWFQEVHDMF-BYPYZUCNSA-N
SMILES
CC(C)[C@H](N)C(O)=O
Appearance (Color)
White
Loss on drying
=<0.3 %
Specific optical rotation
+26.5° to +29.0° (20°C, 589 nm) (c=8, 6 N HCl)
Chloride (Cl)
=<500 ppm
Ammonium (NH4)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text