L-Valine 99%
Thermo Scientific Chemicals

L-Valine 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 140810250
Price (USD)/ Each
39.65
Online exclusive
43.40 
Save 3.75 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
39.65
Online exclusive
43.40 
Save 3.75 (9%)
Add to cart
L-Valine 99%
Catalog number140810250
Price (USD)/ Each
39.65
Online exclusive
43.40 
Save 3.75 (9%)
-
Add to cart
Chemical Identifiers
CAS72-18-4
IUPAC Name(2S)-2-amino-3-methylbutanoic acid
Molecular FormulaC5H11NO2
InChI KeyKZSNJWFQEVHDMF-BYPYZUCNSA-N
SMILESCC(C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Sulfate (SO4)=<300 ppm
Appearance (Color)White
Loss on drying=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only