Vanillin, 99%, pure, Thermo Scientific Chemicals
Vanillin, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanillin, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
2 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 140822500
Price (USD)/ Each
79.65
Online exclusive
87.90 
Save 8.25 (9%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
87.90 
Save 8.25 (9%)
Add to cart
Vanillin, 99%, pure, Thermo Scientific Chemicals
Catalog number140822500
Price (USD)/ Each
79.65
Online exclusive
87.90 
Save 8.25 (9%)
-
Add to cart
Chemical Identifiers
CAS121-33-5
IUPAC Name4-hydroxy-3-methoxybenzaldehyde
Molecular FormulaC8H8O3
InChI KeyMWOOGOJBHIARFG-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point81°C to 84°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only