4-Vinyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals
4-Vinyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Vinyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
Packaging:
Glass bottle
Catalog number 140885000
Price (USD)/ Each
124.00
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
124.00
Add to cart
4-Vinyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals
Catalog number140885000
Price (USD)/ Each
124.00
-
Add to cart
Chemical Identifiers
IUPAC Name4-ethenylcyclohex-1-ene
Molecular FormulaC8H12
InChI KeyBBDKZWKEPDTENS-UHFFFAOYNA-N
SMILESC=CC1CCC=CC1
Molecular Weight (g/mol)108.18
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4620 to 1.4660 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only