Thermo Scientific Chemicals

4-Vinylpyridine, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 140912500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

4-Vinylpyridine, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 140912500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 140912500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-43-6
IUPAC Name
4-ethenylpyridine
Molecular Formula
C7H7N
InChI Key
KFDVPJUYSDEJTH-UHFFFAOYSA-N
SMILES
C=CC1=CC=NC=C1
Appearance (Color)
Clear colorless to yellow to red-brown
Appearance (Form)
Liquid
Appearance
May darken on storage
Infrared spectrum
Conforms
GC
>=94.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text