N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals
N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
500 mL
Catalog number 140920025
Price (USD)/ Each
261.65
Online exclusive
291.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
261.65
Online exclusive
291.00 
Save 29.35 (10%)
Add to cart
N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals
Catalog number140920025
Price (USD)/ Each
261.65
Online exclusive
291.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS88-12-0
IUPAC Name1-ethenylpyrrolidin-2-one
Molecular FormulaC6H9NO
InChI KeyWHNWPMSKXPGLAX-UHFFFAOYSA-N
SMILESC=CN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
GC=<0.2 % (Pyrrolidone)
Refractive index1.5110 to 1.5140 (20°C, 589 nm)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only