1x1 image pixel for data collection
Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, 99%
Thermo Scientific Chemicals

Yohimbine hydrochloride, 99%

Yohimbine hydrochloride, CAS # 65-19-0, is an α2-adrenergic receptor antagonist, which blocks the central α2-adrenegic receptors in the brain. | CAS: 65-19-0 | C21H27ClN2O3 | 390.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 141030050
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS65-19-0
IUPAC Namehydrogen methyl (1S,15R,18S,19R,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
Molecular FormulaC21H27ClN2O3
InChI KeyPIPZGJSEDRMUAW-VJDCAHTMSA-N
SMILES[H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Specific optical rotation+99° to +105° (20°C, 589 nm) (c=1, H2O)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Yohimbine hydrochloride is an indole alkaloid derived from the tree bark
  • It is an antagonist of α2-adrenergic receptors in the brain, reducing the effects of xylazine, which is an α2-adrenergic agonist

Applications

  • Yohimbine hydrochloride blocks the central α2-adrenegic receptors in the brain
  • It can reduce or eradicate effects of xylazine, which is an α2-adrenergic agonist
  • It can have an antidiuretic effect
RUO – Research Use Only

General References:

  1. Zhang, W. and Roomans, G.M. A yohimbine-dependent, UK14,304 induced ion transient in HT29 cells studied by X-ray microanalysis. Scanning microscopy, 1996, 10 (1), 293-299.
  2. Susan, X. Jiang et. al. Reversible inhibition of intracellular calcium influx through NMDA receptors by imidazoline I2receptor antagonists. European Journal of Pharmacology. 2010, 629,(1), 12-19.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link