1x1 image pixel for data collection
HBSS, no calcium, no magnesium
Save up to 29% on your order of cell culture products. View promotion details >
HBSS, no calcium, no magnesium
Gibco™

HBSS, no calcium, no magnesium

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
14170112500 mL
14170120100 mL
1417016110 x 500 mL
Catalog number 14170112
Price (USD)
30.65
Online Exclusive
32.75
Save 2.10 (6%)
Each
-
Add to cart
Quantity:
500 mL
Customize this product
Price (USD)
30.65
Online Exclusive
32.75
Save 2.10 (6%)
Each
Add to cart

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.


This HBSS is modified as follows:
WithWithout
• Phenol Red• Calcium
• Glucose• Magnesium


The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
AdditiveNo Sodium Pyruvate
ClassificationAnimal Origin-Free
Dilution1 X
For Use With (Application)Mammalian Cell Culture
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality235 - 295 mOsm/kg
Phenol Red IndicatorPhenol Red
Product LineGibco™
Quantity500 mL
Shipping ConditionRoom Temperature
SterilitySterile-filtered
FormLiquid
Product TypeHanks' Balanced Salt Solution (HBSS)
Solution TypeHank's Balanced Salt
pH6.7 to 7.8
Unit SizeEach
Contents & Storage
Storage conditions: 15-30° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Please enter sku to load default questions

Frequently asked questions (FAQs)

Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?

The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link