DPBS, no calcium, no magnesium
Love your cells 프로모션에 해당하는 제품으로 프로모션 페이지에서 할인된 가격을 만나보세요!
DPBS, no calcium, no magnesium
Gibco™

DPBS, no calcium, no magnesium

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging Type
1419025010 x 500 mLBottle
14190094
also known as 14190-094
500 mLBottle
14190144500 mLBottle
141901361000 mLBottle
141902356 x 1000 mLBottle
141903425 LBag
1419035910 LBag
1419036720 LBag
Catalog number 14190250
Price (KRW)
317,000
Online offer
Ends: 30-Jun-2025
333,000
Save 16,000 (5%)
Each
Add to cart
Quantity:
10 x 500 mL
Packaging Type:
Bottle
Customize this product
Price (KRW)
317,000
Online offer
Ends: 30-Jun-2025
333,000
Save 16,000 (5%)
Each
Add to cart

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
AdditiveNo Sodium Pyruvate
ClassificationAnimal Origin-Free
ColorClear
Dilution1 X
For Use With (Application)Mammalian Cell Culture
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 - 300 mOsm/kg
Packaging TypeBottle
Phenol Red IndicatorNo Phenol Red
Product LineGibco™
Quantity10 x 500 mL
Shipping ConditionRoom Temperature
SterilitySterile-filtered
FormLiquid
Product TypeDPBS
Solution TypeDulbecco's Phosphate Buffered Saline
pH7.0 to 7.3
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture

Share catalog number, name or link