Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 340 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 375 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 405 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 414 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan |
Wavelength | 450 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 492 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 520 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 530 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 540 nm |
Unit Size | Each |
Product Type | Optical Filter |
For Use With (Equipment) | Multiskan FC |
Product Line | Multiskan™ |
Wavelength | 550 nm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Product Type | Wavelength | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1423405 | Each | Optical Filter | 340 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1423755 | Each | Optical Filter | 375 nm | 1,266.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1424055 | Each | Optical Filter | 405 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1424145 | Each | Optical Filter | 414 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1424505 | Each | Optical Filter | 450 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1424925 | Each | Optical Filter | 492 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1425205 | Each | Optical Filter | 520 nm | 1,298.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1425305 | Each | Optical Filter | 530 nm | 1,666.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1425405 | Each | Optical Filter | 540 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1425505 | Each | Optical Filter | 550 nm | 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|