Dynabeads™ Antibody Coupling Kit
Dynabeads™ Antibody Coupling Kit
Invitrogen™

Dynabeads™ Antibody Coupling Kit

The Dynabeads™ Antibody Coupling Kit is intended for the coupling of an antibody or protein to the surface of Dynabeads™Read more
Have Questions?
Catalog NumberQuantity
14311D60 mg
Catalog number 14311D
Price (EUR)
534,00
Each
-
Add to cart
Quantity:
60 mg
Recurring order eligible. Learn more »
Price (EUR)
534,00
Each
Add to cart
Ask our AI about this Product

The Dynabeads™ Antibody Coupling Kit is intended for the coupling of an antibody or protein to the surface of Dynabeads™ M-270 Epoxy beads. The coupled beads and supplied buffers can then be used for experiments such as immunoassays, immunoprecipitation (IP), Co-IP of protein complexes, chromatin IP (ChIP), RNA-IP (RIP), cell isolation, etc. The Dynabeads Epoxy beads in this kit do not contain Tween™ detergent making them suitable for mass spectrometry (MS) analysis. The kit includes the buffers needed for efficient antibody coupling and washing steps. Note that the SB buffer supplied in the kit does contain Tween™ detergent, which means the buffer will need to be replaced with standard TBS or PBS buffer if the kit is to be used for MS.

Benefits of Dynabeads Antibody Coupling Kit include:
Convenience—contains optimized buffers and epoxy-activated Dynabeads magnetic beads
Efficiency­—easy-to-use protocol with minimal hands-on time
Flexibility­—allows coupling of any antibody of your choice
Robustness—covalent antibody coupling to the beads avoids co-elution of the antibody with the target proteins

Efficient and easy coupling
With this kit antibodies are covalently coupled to Dynabeads M-270 Epoxy beads, minimizing the risk of bound antibodies contaminating the final eluate. Dynabeads M-270 Epoxy beads exhibit extremely low background binding and do not require blocking before use. Other proteins such as lectins and enzymes may also be coupled using the same beads, buffers, and protocol (depending on the stability and functionality of the protein/ligand you are using).

Magnetic bead-based separation offers easy handling
Captured proteins and protein complexes are easily separated, washed, and eluted using a DynaMag magnet due to the magnetic properties of the Dynabeads M-270 Epoxy beads. The supplied buffers are optimized to give consistent and reliable results. In addition to Dynabeads M-270 Epoxy beads, the kit includes five different binding, washing, and elution buffers. Target specific antibody is required for coupling, but not included.

Automation-ready for use with KingFisher purification systems
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers.

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Antibody Coupling Kit in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM page.

Learn more about Dynabeads magnetic beads and KingFisher Sample Purification Systems:
Dynabeads M-270 Epoxy beads supplied in the kit can be purchased separately
See our immunoprecipitation selection table, data, and references
Dynabeads magnetic beads selection guide
See magnets for Dynabeads magnetic beads separations
KingFisher automation protocols
Watch a video about the KingFisher Flex instrument

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceISO9001 and ISO13485
ColorBrown
DescriptionFor immoblization of antibodies to magnetic beads
Diameter (Metric)2.8 μm
Final Product TypeAntibody (or other protein ligand) coupled Dynabeads
For Use With (Application)Immunoprecipitation
For Use With (Equipment)KingFisher™ Sample Purification Systems, DynaMag™ Magnets
FormatFreeze-dried beads
High-throughput CompatibilityHigh-throughput Compatible
MaterialPolystyrene
Quantity60 mg
Regulatory StatusFor Research Use Only
Sample TypeIntact Proteins (antibodies, purified antibodies, peptides, functional enzymes)
Shelf Life24 months from date of manufacture
Shipping ConditionAmbient temperature
Surface FunctionalityEpoxy
UniformityMonosized 2.8 μm (CV <5%)
TypeAntibody Coupling Kit
Unit SizeEach
Contents & Storage
Dynabeads M-270 Epoxy beads (60 mg) and 5 different buffers for covalent coupling and washing Store at 2°C to 8°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Videos

Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Customers who viewed this item also viewed



      Documents & Downloads

      Certificates

      Lot #Certificate TypeDateCatalog Number(s)
      3257695Certificate of AnalysisJun 10, 202514311D
      3249509Certificate of AnalysisMay 29, 202514311D
      3247895Certificate of AnalysisMay 27, 202514311D
      3247883Certificate of AnalysisMay 27, 202514311D
      3243889Certificate of AnalysisMay 21, 202514311D
      5 results displayed, search above for a specific certificate

      Safety Data Sheets

      Frequently asked questions (FAQs)

      Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

      - The solution is too viscous.
      - The beads have formed aggregates because of protein-protein interaction.

      Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
      - Add DNase I to the lysate (~0.01 mg/mL)
      - Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
      - Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

      Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

      For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

      Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

      Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

      Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

      Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

      Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

      There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

      For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

      Protocol:

      1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
      2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
      3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
      4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

      For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

      Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

      Protocol:

      1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
      2.Pipette out desired amount of human IgG in a separate tube.
      3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
      4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
      5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
      6.Wash the beads (with coating) four times, and measure cpm again.
      The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

      Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

      Other products to consider



      Share catalog number, name or link

      1x1 image pixel for data collection