Nunc™ IVF Multi Dish, 4-well
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ IVF Multi Dish, 4-well
Thermo Scientific™

Nunc™ IVF Multi Dish, 4-well

Perform applications which yield consistently reliable, high-quality results during vitrification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface Treatment
144444Nunclon Delta
179830Non-treated
Catalog number 144444
Price (USD)
619.65
Online Exclusive
668.00
Save 48.35 (7%)
Case of 120
-
Add to cart
Surface Treatment:
Nunclon Delta
Price (USD)
619.65
Online Exclusive
668.00
Save 48.35 (7%)
Case of 120
Add to cart

Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.

Innovative features for high quality and ease-of-use:

  • Ensures proper well orientation and prevents cross contamination with notched lid and dish
  • Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
  • The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
  • The non-treated hydrophobic surface facilitates microdrop embryo culture
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
DescriptionNunclon Delta-treated IVF Dish, 4-well
Length (Metric)66 mm
No. per Case120
No. per Pack4
Width (Metric)66 mm
Culture Area1.9 cm2/well
MaterialPolystyrene (USP Class VI)
SterilitySterile
Surface TreatmentNunclon Delta
Well Count4
Unit SizeCase of 120

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link