Thermo Scientific Chemicals

(Methoxymethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 146011000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

(Methoxymethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 146011000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 146011000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4009-98-7
IUPAC Name
(methoxymethyl)triphenylphosphanium chloride
Molecular Formula
C20H20ClOP
InChI Key
SJFNDMHZXCUXSA-UHFFFAOYSA-M
SMILES
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
HPLC
>=97.5 %
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Water
=<0.5 %
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text