(Methoxymethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
(Methoxymethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Methoxymethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 146015000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS4009-98-7
IUPAC Name(methoxymethyl)triphenylphosphanium chloride
Molecular FormulaC20H20ClOP
InChI KeySJFNDMHZXCUXSA-UHFFFAOYSA-M
SMILES[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Water=<0.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only