Thermo Scientific Chemicals

4,4'-Dimethoxytrityl Chloride, 98%, Thermo Scientific Chemicals

Catalog number: 146020250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4,4'-Dimethoxytrityl Chloride, 98%, Thermo Scientific Chemicals

Catalog number: 146020250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 146020250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40615-36-9
IUPAC Name
1-[chloro(4-methoxyphenyl)phenylmethyl]-4-methoxybenzene
Molecular Formula
C21H19ClO2
InChI Key
JBWYRBLDOOOJEU-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Titration Argentometric
97.5 to 102.5 % (ex Cl)
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
121°C to 127°C
Appearance (Color)
Pink to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text