Thermo Scientific Chemicals

(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, balance TPPO, Thermo Scientific Chemicals

Catalog number: 146080050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, balance TPPO, Thermo Scientific Chemicals

Catalog number: 146080050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 146080050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5717-37-3
IUPAC Name
ethyl 2-(triphenyl-λ⁵-phosphanylidene)propanoate
Molecular Formula
C23H23O2P
InChI Key
KZENFXVDPUMQOE-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
HPLC
ca. 94 %
Infrared spectrum
Conforms
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text