3-Aminophenol, 99%, Thermo Scientific Chemicals
3-Aminophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminophenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number 146102500
Price (USD)/ Each
66.65
Online exclusive
78.30 
Save 11.65 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
78.30 
Save 11.65 (15%)
Add to cart
3-Aminophenol, 99%, Thermo Scientific Chemicals
Catalog number146102500
Price (USD)/ Each
66.65
Online exclusive
78.30 
Save 11.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-aminophenol
Molecular FormulaC6H7NO
InChI KeyCWLKGDAVCFYWJK-UHFFFAOYSA-N
SMILESNC1=CC=CC(O)=C1
Molecular Weight (g/mol)109.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Melting point120°C to 124°C
GC>=98.5 %
Appearance (Color)White to grey to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only